Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200008312.mm.1 |
NM_001198872.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008312.mm.1 |
NM_001198872.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008312.mm.1 |
NM_001198873.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008312.mm.1 |
NM_001198873.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008312.mm.1 |
NM_001198877.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008312.mm.1 |
NM_001198877.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008312.mm.1 |
NM_001198878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008312.mm.1 |
NM_001198878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008312.mm.1 |
TC0200001033.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
NM_001198872.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
NM_001198872.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
NM_001198873.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
NM_001198873.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
NM_001198874.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
NM_001198874.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
NM_001198877.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
NM_001198877.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
NM_001198878.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
NM_001198878.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008319.mm.1 |
TC0200001033.mm.1 |
MTA TC |