Assay Details
| Gene Symbol: Gpcpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042672.1 | NP_001036137.1 | 17 - 18 | 1748 | 143 | |
| NM_001291050.1 | NP_001277979.1 | 16 - 17 | 1621 | 143 | ||
| NM_001291051.1 | NP_001277980.1 | 16 - 17 | 1651 | 143 | ||
| NM_001291052.1 | NP_001277981.1 | 16 - 17 | 1669 | 143 | ||
| NM_027096.2 | NP_081372.1 | 15 - 16 | 1545 | 143 | ||
| NM_028802.3 | NP_083078.3 | 17 - 18 | 1745 | 143 | ||
| GenBank mRNA | AK009563.1 | - | 10 - 11 | 964 | 143 | |
| AK030645.1 | - | 17 - 18 | 1699 | 143 | ||
| AK031658.1 | - | 16 - 17 | 1624 | 143 | ||
| AK050318.1 | - | 17 - 18 | 1660 | 143 | ||
| AK051728.1 | - | 9 - 10 | 808 | 143 | ||
| AK122510.1 | - | 4 - 5 | 2400 | 143 | ||
| AK141374.1 | - | 17 - 18 | 1635 | 143 | ||
| AK163419.1 | - | 16 - 17 | 1592 | 143 | ||
| AK165952.1 | - | 17 - 18 | 1712 | 143 | ||
| AK166293.1 | - | 8 - 9 | 679 | 143 | ||
| AK166454.1 | - | 16 - 17 | 1634 | 143 | ||
| BC006887.1 | - | 16 - 17 | 1593 | 143 | ||
| BC033408.1 | - | 15 - 16 | 1475 | 5357683 | 143 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpcpd1
Genomic Map