Assay Details
| Gene Symbol: Ssfa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080558.4 | NP_542125.3 | 4 - 5 | 471 | 65 | |
| GenBank mRNA | AB093303.1 | - | 4 - 5 | 346 | 65 | |
| AB120565.1 | - | 4 - 5 | 356 | 65 | ||
| AK037213.1 | - | 2 - 3 | 181 | 65 | ||
| AK169543.1 | - | 4 - 5 | 472 | 65 | ||
| BC122519.1 | - | 4 - 5 | 357 | 65 | ||
| BC141883.1 | - | 4 - 5 | 356 | 40097831 | 65 | |
| CF539296.1 | - | 4 - 5 | 287 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037213.1 transcripts.
Set Membership: |
|

Ssfa2
Genomic Map