Assay Details
Gene Symbol: Pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244198.1 | NP_001231127.1 | 1 - 2 | 156 | 72 | |
NM_001244201.1 | NP_001231130.1 | 1 - 2 | 156 | 72 | ||
GenBank mRNA | AF457142.1 | - | 1 - 3 | 122 | 72 | |
AK028059.1 | - | 1 - 2 | 157 | 72 | ||
AK032616.1 | - | 1 - 2 | 157 | 72 | ||
BC036957.1 | - | 1 - 2 | 152 | 4504106 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF457142.1, AK028059.1, BC036957.1, NM_001244198.1, NM_001244201.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
