Assay Details
Gene Symbol: Pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244198.1 | NP_001231127.1 | 9 - 10 | 1092 | 79 | |
NM_001244200.1 | NP_001231129.1 | 10 - 11 | 1304 | 79 | ||
NM_001244201.1 | NP_001231130.1 | 8 - 9 | 1050 | 79 | ||
NM_001244202.1 | NP_001231131.1 | 8 - 9 | 1606 | 79 | ||
NM_013627.5 | NP_038655.1 | 9 - 10 | 1648 | 79 | ||
GenBank mRNA | AF443223.1 | - | 5 - 6 | 705 | 79 | |
AF457141.1 | - | 3 - 4 | 371 | 79 | ||
AJ292077.1 | - | 8 - 9 | 909 | 79 | ||
AJ307468.1 | - | 8 - 9 | 916 | 79 | ||
AK028059.1 | - | 8 - 9 | 1051 | 79 | ||
AK032616.1 | - | 9 - 10 | 1093 | 79 | ||
AK045805.1 | - | 8 - 9 | 1608 | 79 | ||
AK139054.1 | - | 636 | 79 | |||
AY064175.1 | - | 4 - 5 | 515 | 79 | ||
BC011272.1 | - | 9 - 10 | 1336 | 4008490 | 79 | |
BC036957.1 | - | 8 - 9 | 1046 | 4504106 | 79 | |
BC069912.1 | - | 5 - 6 | 1083 | 79 | ||
M77842.1 | - | 4 - 5 | 635 | 79 | ||
X63963.1 | - | 9 - 10 | 969 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
