Assay Details
Gene Symbol: Cat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY040626.1 | - | 1 - 2 | 122 | 106 | |
M62897.1 | - | 1 - 2 | 66 | 106 | ||
X52108.1 | - | 1 - 2 | 66 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
