Assay Details
| Gene Symbol: Papola | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011112.3 | NP_035242.1 | 19 - 20 | 2193 | 92 | |
| GenBank mRNA | AK028940.1 | - | 19 - 20 | 2191 | 92 | |
| AK090096.1 | - | 19 - 20 | 2194 | 92 | ||
| AK132704.1 | - | 19 - 20 | 2180 | 92 | ||
| BC060128.1 | - | 18 - 19 | 2040 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Papola
Genomic Map