Assay Details
| Gene Symbol: Dcaf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165980.1 | NP_001159452.1 | 5 - 6 | 786 | 68 | |
| NM_001165981.1 | NP_001159453.1 | 5 - 6 | 786 | 68 | ||
| NM_001165982.1 | NP_001159454.1 | 5 - 6 | 786 | 68 | ||
| NM_198005.2 | NP_932122.2 | 5 - 6 | 786 | 68 | ||
| GenBank mRNA | AK014717.1 | - | 5 - 6 | 750 | 68 | |
| AK032671.1 | - | 5 - 6 | 748 | 68 | ||
| AK079462.1 | - | 5 - 6 | 580 | 68 | ||
| AK081839.1 | - | 5 - 6 | 785 | 68 | ||
| AK160679.1 | - | 5 - 6 | 753 | 68 | ||
| BC031546.1 | - | 5 - 6 | 678 | 3669736 | 68 | |
More Information
Set Membership: |
|

Dcaf17
Genomic Map