Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00008914.mm.1 |
NM_001199357.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008914.mm.1 |
NM_001199357.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008914.mm.1 |
NM_001199360.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008914.mm.1 |
NM_001199360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008914.mm.1 |
NM_177592.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008914.mm.1 |
NM_177592.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008914.mm.1 |
TC0X00001407.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00008909.mm.1 |
NM_001199357.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008909.mm.1 |
NM_001199357.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008909.mm.1 |
NM_177592.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008909.mm.1 |
NM_177592.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00008909.mm.1 |
TC0X00001407.mm.1 |
MTA TC |