Assay Details
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110222.1 | NP_001103692.1 | 4 - 5 | 1059 | 124 | |
NM_001110223.1 | NP_001103693.1 | 4 - 5 | 981 | 124 | ||
NM_001110224.1 | NP_001103694.1 | 4 - 5 | 981 | 124 | ||
NM_010025.2 | NP_034155.2 | 4 - 5 | 981 | 124 | ||
GenBank mRNA | AB011678.1 | - | 4 - 5 | 1077 | 124 | |
AF045547.1 | - | 4 - 5 | 862 | 124 | ||
AK014264.1 | - | 4 - 5 | 982 | 124 | ||
AK048410.1 | - | 4 - 5 | 987 | 124 | ||
AK049050.1 | - | 4 - 5 | 1055 | 124 | ||
AK164119.1 | - | 4 - 5 | 868 | 124 | ||
AY560329.1 | - | 4 - 5 | 869 | 124 | ||
BC056391.1 | - | 4 - 5 | 874 | 6855006 | 124 | |
BC057010.1 | - | 4 - 5 | 966 | 6825750 | 124 | |
BC062974.1 | - | 4 - 5 | 966 | 6825750 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
