Assay Details
Gene Symbol: Slc13a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004148.4 | NP_001004148.1 | 6 - 7 | 880 | 94 | |
GenBank mRNA | AK043499.1 | - | 6 - 7 | 881 | 94 | |
AK048662.1 | - | 6 - 7 | 881 | 94 | ||
AK140943.1 | - | 6 - 7 | 882 | 94 | ||
AK158253.1 | - | 6 - 7 | 884 | 94 | ||
AY368893.1 | - | 6 - 7 | 841 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
