Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200031987.mm.1 |
NM_001018042.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031987.mm.1 |
NM_001018042.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031987.mm.1 |
NM_001098425.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031987.mm.1 |
NM_001098425.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031987.mm.1 |
TC0200003745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031986.mm.1 |
NM_001018042.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031986.mm.1 |
NM_001018042.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031986.mm.1 |
NM_001098425.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031986.mm.1 |
NM_001098425.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031986.mm.1 |
TC0200003745.mm.1 |
MTA TC |