Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200031968.mm.1 |
NR_102348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031968.mm.1 |
NR_102349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031968.mm.1 |
TC0200003744.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031966.mm.1 |
NR_102348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031966.mm.1 |
NR_102349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031966.mm.1 |
TC0200003744.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031967.mm.1 |
NR_102348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031967.mm.1 |
NR_102349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031967.mm.1 |
TC0200003744.mm.1 |
MTA TC |