Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200033451.mm.1 |
NM_001085448.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033451.mm.1 |
NM_001085448.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033451.mm.1 |
NM_001085449.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033451.mm.1 |
NM_001085449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033451.mm.1 |
NM_001085450.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033451.mm.1 |
NM_001085450.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033451.mm.1 |
NM_001085453.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033451.mm.1 |
NM_007615.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033451.mm.1 |
NM_007615.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033451.mm.1 |
TC0200003869.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200033453.mm.1 |
NM_001085448.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033453.mm.1 |
NM_001085448.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033453.mm.1 |
NM_001085449.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033453.mm.1 |
NM_001085449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033453.mm.1 |
NM_001085450.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033453.mm.1 |
NM_001085450.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033453.mm.1 |
NM_001085453.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033453.mm.1 |
NM_007615.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033453.mm.1 |
NM_007615.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033453.mm.1 |
TC0200003869.mm.1 |
MTA TC |