Assay Details
Gene Symbol: Trappc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024206.2 | NP_001019377.1 | 2 - 3 | 180 | 72 | |
GenBank mRNA | BC115818.1 | - | 1 - 2 | 149 | 40107010 | 72 |
BC115921.1 | - | 1 - 2 | 149 | 40107011 | 72 | |
CK621128.1 | - | 2 - 3 | 180 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
