Assay Details
Gene Symbol: Trappc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024206.2 | NP_001019377.1 | 4 - 5 | 393 | 73 | |
GenBank mRNA | BC115818.1 | - | 3 - 4 | 362 | 40107010 | 73 |
BC115921.1 | - | 3 - 4 | 362 | 40107011 | 73 | |
BY479576.1 | - | 2 - 3 | 254 | 73 | ||
CK621128.1 | - | 4 - 5 | 393 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27894315 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054883 | BC115921.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054883 | BC115818.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028310 | CK621128.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028310 | NM_001024206.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054887 | BY479576.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054887 | BC115921.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054887 | NM_001024206.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054887 | BC115818.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850718 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 179974 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054887 | BY479576.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054887 | BC115921.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054887 | NM_001024206.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054887 | BC115818.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028310 | CK621128.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028310 | NM_001024206.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894316 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894321 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894323 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894317 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894318 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894320 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894315 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894328 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894324 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27894329 | - | - |
Whole Mouse Genome G4122A | A_52_P298867 | BY479576.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P298867 | CK621128.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P298867 | BC115921.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P298867 | NM_001024206.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P298867 | BC115818.1 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09330640 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09224188 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09224190 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09330628 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09330613 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09224194 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09224185 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27894315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894364 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894343 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894331 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894338 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27894389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27894315 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09224176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27894320 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27894321 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27894316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27894328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27894329 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09224169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09224185 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27894325 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10377512 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10377513 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10377510 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10377511 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10377509 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10377514 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10377515 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436019_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10377508 | - | - |
Mouse Genome 430 2.0 Array | 1436019_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1436019_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245828_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245828_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100007680.mm.1 | NM_001024206.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100007680.mm.1 | NM_001024206.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100007680.mm.1 | TC1100000907.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100007681.mm.1 | NM_001024206.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100007681.mm.1 | NM_001024206.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100007681.mm.1 | TC1100000907.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100007682.mm.1 | NM_001024206.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100007682.mm.1 | NM_001024206.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100007682.mm.1 | TC1100000907.mm.1 | MTA TC |
Mouse Expression Set 430 | 1436019_a_at | - | - |
Mouse Expression Set 430 | 1436019_PM_a_at | - | - |
Murine Genome U74v2 Set | 95629_PM_at | - | - |
Murine Genome U74v2 Set | 95629_at | - | - |
Mouse Genome 430A 2.0 Array | 1436019_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1436019_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1436019_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1436019_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 657074 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1213711 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2943661 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2943661 | - | - |
