Assay Details
Gene Symbol: Galnt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173030.2 | NP_766618.2 | 10 - 11 | 1807 | 78 | |
GenBank mRNA | AB082928.1 | - | 8 - 9 | 1295 | 78 | |
AK030726.1 | - | 9 - 10 | 1739 | 78 | ||
AK038387.1 | - | 10 - 11 | 1808 | 78 | ||
AK045417.1 | - | 10 - 11 | 1808 | 78 | ||
BC131651.1 | - | 10 - 11 | 1486 | 40110517 | 78 | |
BC131652.1 | - | 9 - 10 | 1517 | 40110518 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
