Assay Details
Gene Symbol: Etl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK052020.1 | - | 2 - 3 | 588 | 121 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK052020.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
