Assay Details
| Gene Symbol: Gpat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081089.2 | NP_001074558.2 | 20 - 21 | 2320 | 62 | |
| GenBank mRNA | AB257502.1 | - | 19 - 20 | 2238 | 62 | |
| AK040991.1 | - | 6 - 7 | 786 | 62 | ||
| AK131154.1 | - | 20 - 21 | 2319 | 62 | ||
| BC113165.1 | - | 20 - 21 | 2314 | 40059976 | 62 | |
| BC113776.1 | - | 20 - 21 | 2311 | 40059969 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpat2
Genomic Map