Assay Details
| Gene Symbol: Ighg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037822.1 | - | 731 | 86 | ||
| AK131185.1 | - | 918 | 86 | |||
| AY571287.1 | - | 775 | 86 | |||
| BC055910.1 | - | 802 | 86 | |||
| D14625.1 | - | 797 | 86 | |||
| L36938.1 | - | 2 - 3 | 333 | 86 | ||
More Information
Set Membership: |
|

Ighg3
Genomic Map