Assay Details
| Gene Symbol: Xirp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024618.2 | NP_001019789.1 | 1 - 2 | 152 | 66 | |
| NM_001083919.1 | NP_001077388.1 | 1 - 2 | 152 | 66 | ||
| GenBank mRNA | AK040786.1 | - | 1 - 2 | 153 | 66 | |
| DQ011666.1 | - | 1 - 2 | 153 | 66 | ||
| EF119712.1 | - | 1 - 2 | 152 | 66 | ||
| EF119716.1 | - | 1 - 2 | 152 | 66 | ||
| EU286531.1 | - | 1 - 2 | 135 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK040786.1, DQ011666.1, EF119712.1, EF119716.1, EU286531.1, NM_001024618.2, NM_001083919.1 transcripts.
Set Membership: |
|

Xirp2
Genomic Map