Assay Details
| Gene Symbol: Xirp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024618.2 | NP_001019789.1 | 3 - 4 | 436 | 137 | |
| NM_001083919.1 | NP_001077388.1 | 3 - 4 | 436 | 137 | ||
| GenBank mRNA | AK040786.1 | - | 3 - 4 | 437 | 137 | |
| AY775570.1 | - | 2 - 3 | 290 | 137 | ||
| DQ011666.1 | - | 3 - 4 | 437 | 137 | ||
| EF119712.1 | - | 3 - 4 | 436 | 137 | ||
| EF119713.1 | - | 592 | 137 | |||
| EF119714.1 | - | 640 | 137 | |||
| EF119715.1 | - | 667 | 137 | |||
| EF119716.1 | - | 3 - 4 | 436 | 137 | ||
| EF122140.1 | - | 2 - 3 | 191 | 137 | ||
| KM273012.1 | - | 878 | 137 | |||
More Information
Set Membership: |
|

Xirp2
Genomic Map