Assay Details
| Gene Symbol: 4930517M08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015816.1 | - | 2 - 3 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930548J01 gene | 4930548J01Rik | 1700113K09Rik | -513.7kb | Anti-sense | Chr. 17 - 4119446 - 4122102 on NCBI Build GRCm38 |
| Downstream | AT rich interactive domain 1B (SWI-like) | Arid1b |
8030481M12, 9330189K18Rik, AI836955, Ardi1b, B230217J03Rik, mKIAA1235 |
355.3kb | Sense | Chr. 17 - 4992755 - 5347656 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930517M08Rik
Genomic Map