Assay Details
| Gene Symbol: Gm13481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042510.1 | - | 308 | 143 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream |
predicted gene, 31324 |
Gm31324 | 483.1kb | Anti-sense | Chr. 2 - 48793307 - 48798902 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm13481
Genomic Map