Assay Details
| Gene Symbol: Gm13481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042510.1 | - | 484 | 149 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700019E08 gene | 1700019E08Rik | -2.4mb | Anti-sense | Chr. 2 - 45696605 - 45698447 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31324 |
Gm31324 | 706.1kb | Anti-sense | Chr. 2 - 48793307 - 48798902 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm13481
Genomic Map