Assay Details
| Gene Symbol: 4930500J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040322.1 | 3 - 4 | 349 | 77 | ||
| GenBank mRNA | AK015665.1 | - | 3 - 4 | 351 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

4930500J02Rik
Genomic Map