Assay Details
| Gene Symbol: 9430041J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033568.1 | 6 - 7 | 1436 | 86 | ||
| GenBank mRNA | AK051461.1 | - | 5 - 6 | 1024 | 86 | |
| AK082200.1 | - | 6 - 7 | 1438 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leukocyte-associated Ig-like receptor 1 | Lair1 |
5133400O11Rik, BB115266, D7Bwg0421e, Lair-1 |
-2.7kb | Sense | Chr. 7 - 4007073 - 4073507 on NCBI Build GRCm38 |
| Downstream | tweety homolog 1 (Drosophila) | Ttyh1 |
4930459B04Rik, 6330408P11Rik, TTY1, mTTY1, tty |
23.9kb | Anti-sense | Chr. 7 - 4119527 - 4136241 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2236228 | AK051461.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514675 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689722 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367267 | AK031273.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367267 | AK047517.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482589 | AF343565.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482589 | AK006111.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482589 | AK131722.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482589 | AK138562.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482589 | BC049625.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482589 | NM_080637.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226265 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141410 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639860 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916278 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730931 | AK012332.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486688 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906804 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 401618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 46719 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 31046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85025 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173085 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171562 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2236228 | AK051461.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06113056 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171562 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01475124 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01475155 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173085 | - | - |
| Whole Mouse Genome G4122A | A_52_P35202 | AK020464.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P35202 | AK051461.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P35202 | AK082200.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P35202 | NR_033568.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125465 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128008 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173085 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01475155 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06113056 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06132296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24947626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24947625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24947629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06116768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24947665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06132294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24947661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24947627 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559499 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453671_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559498 | - | - |
| Mouse Genome 430 2.0 Array | 1453671_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453671_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700019497.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019497.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019497.mm.1 | NR_033568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019501.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019501.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019501.mm.1 | NR_033568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019502.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019502.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019502.mm.1 | NR_033568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019508.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019508.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019508.mm.1 | NR_033568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019504.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019504.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019504.mm.1 | NR_033568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019507.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019507.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019507.mm.1 | NR_033568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019490.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019490.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019490.mm.1 | NR_033568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019494.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019494.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019494.mm.1 | NR_033568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019493.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019493.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019493.mm.1 | NR_033568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019495.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019495.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019495.mm.1 | NR_033568.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019505.mm.1 | TC0700002145.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019505.mm.1 | NR_033568.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019505.mm.1 | NR_033568.1 | Refseq |
| Mouse Expression Set 430 | 1453671_at | - | - |
| Mouse Expression Set 430 | 1453671_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453671_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453671_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 723416 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104150092 | - | - |

9430041J12Rik
Genomic Map