Assay Details
Gene Symbol: Gm40918 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC048601.1 | - | 98 | 105 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | expressed sequence AI463229 | AI463229 | 9130430E04 | -47.3kb | Sense | Chr. 13 - 37657013 - 37670077 on NCBI Build GRCm38 |
Downstream | ras responsive element binding protein 1 | Rreb1 |
1110037N09Rik, AA414966, B930013M22Rik |
60.7kb | Anti-sense | Chr. 13 - 37778382 - 37952005 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
