Assay Details
Gene Symbol: Gm40918 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC048601.1 | - | 312 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | expressed sequence AI463229 | AI463229 | 9130430E04 | -30.8kb | Sense | Chr. 13 - 37657013 - 37670077 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
