Assay Details
| Gene Symbol: Stkld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198628.2 | NP_941030.2 | 16 - 17 | 1712 | 63 | |
| GenBank mRNA | BC050806.1 | - | 16 - 17 | 1713 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stkld1
Genomic Map