Assay Details
| Gene Symbol: Pdlim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114088.2 | NP_001107560.1 | 10 - 11 | 1135 | 64 | |
| GenBank mRNA | AK010339.1 | - | 8 - 9 | 1128 | 64 | |
| AK128987.1 | - | 9 - 10 | 2087 | 64 | ||
| AK142933.1 | - | 10 - 11 | 1123 | 64 | ||
| AK167476.1 | - | 10 - 11 | 1127 | 64 | ||
| BC038175.1 | - | 3 - 4 | 64 | |||
| BC045528.1 | - | 1 - 2 | 134 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdlim7
Genomic Map