Assay Details
| Gene Symbol: Fcnaos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039020.1 | - | 5 - 6 | 962 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 141 | Tmem141 |
1110065P14Rik, AI663999, D2Ertd217e |
-5.6kb | Anti-sense | Chr. 2 - 25620066 - 25622039 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 35630 |
Gm35630 | 6.4kb | Sense | Chr. 2 - 25635518 - 25640293 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Fcnaos
Genomic Map