| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200008368.mm.1 |
NR_028265.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200008368.mm.1 |
NR_028265.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200008368.mm.1 |
TC1200001192.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200008371.mm.1 |
NR_028265.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200008371.mm.1 |
NR_028265.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200008371.mm.1 |
TC1200001192.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200008370.mm.1 |
NR_028265.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200008370.mm.1 |
NR_028265.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200008370.mm.1 |
TC1200001192.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200008380.mm.1 |
NR_028265.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200008380.mm.1 |
NR_028265.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200008380.mm.1 |
TC1200001192.mm.1 |
MTA TC |