Assay Details
| Gene Symbol: Gm26688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125899.1 | 2 - 3 | 429 | 104 | ||
| GenBank mRNA | AK017409.1 | - | 2 - 3 | 430 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
transcription factor AP-2, alpha |
Tfap2a |
AP-2, AP2alpha, Ap-2 (a), Ap2, Ap2tf, Tcfap2a |
Anti-sense | Chr. 13 - 40713802 - 40738238 on NCBI Build GRCm38 | |
| Downstream |
glucosaminyl (N-acetyl) transferase 2, I-branching enzyme |
Gcnt2 |
5330430K10Rik, IGnT, IGnTA, IGnTB, IGnTC |
124.6kb | Sense | Chr. 13 - 40859768 - 40960892 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26688
Genomic Map