Assay Details
| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271879.1 | NP_001258808.1 | 9 - 10 | 1153 | 100 | |
| NM_177694.6 | NP_808362.2 | 10 - 11 | 1195 | 100 | ||
| NR_073508.1 | 10 - 11 | 1188 | 100 | |||
| GenBank mRNA | AB125740.1 | - | 10 - 11 | 1195 | 100 | |
| AB206762.1 | - | 10 - 11 | 1195 | 100 | ||
| AK034197.1 | - | 10 - 11 | 1177 | 100 | ||
| BC109163.2 | - | 9 - 10 | 1075 | 40047987 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ano5
Genomic Map