Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300011721.mm.1 |
NM_019930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011721.mm.1 |
NM_019930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011721.mm.1 |
TC1300001882.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011719.mm.1 |
NM_019930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011719.mm.1 |
NM_019930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011719.mm.1 |
TC1300001882.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011720.mm.1 |
NM_019930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011720.mm.1 |
NM_019930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011720.mm.1 |
TC1300001882.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011718.mm.1 |
NM_019930.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011718.mm.1 |
NM_019930.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011718.mm.1 |
TC1300001882.mm.1 |
MTA TC |