Assay Details
Gene Symbol: Ranbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019930.2 | NP_064314.2 | 7 - 8 | 1219 | 98 | |
GenBank mRNA | AF006465.1 | - | 8 - 9 | 1048 | 98 | |
AK132714.1 | - | 7 - 8 | 698 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
