Assay Details
Gene Symbol: Ippk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199056.3 | NP_951011.1 | 1 - 2 | 244 | 75 | |
GenBank mRNA | AK160092.1 | - | 1 - 2 | 191 | 75 | |
AK167467.1 | - | 1 - 2 | 219 | 75 | ||
BB619387.1 | - | 1 - 2 | 220 | 75 | ||
BC062167.1 | - | 1 - 2 | 156 | 6517021 | 75 | |
BC065157.1 | - | 1 - 2 | 159 | 6416865 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
