Assay Details
Gene Symbol: Pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012076.1 | - | 1 - 2 | 101 | 105 | |
AK075604.1 | - | 1 - 2 | 100 | 105 | ||
AK135948.1 | - | 1 - 2 | 101 | 105 | ||
BB868274.1 | - | 1 - 2 | 161 | 105 | ||
BC003793.1 | - | 1 - 2 | 68 | 3601675 | 105 | |
U28016.1 | - | 1 - 2 | 77 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK012076.1, BC003793.1, U28016.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
