Assay Details
| Gene Symbol: Trdmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010067.4 | NP_034197.3 | 4 - 5 | 353 | 128 | |
| GenBank mRNA | AF012129.1 | - | 4 - 5 | 340 | 128 | |
| AF045889.1 | - | 4 - 5 | 345 | 128 | ||
| AK029136.1 | - | 4 - 5 | 354 | 128 | ||
| AK050403.1 | - | 4 - 5 | 353 | 128 | ||
| BC087892.1 | - | 4 - 5 | 353 | 30504522 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trdmt1
Genomic Map