Assay Details
Gene Symbol: Anapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008569.2 | NP_032595.2 | 15 - 16 | 2049 | 61 | |
GenBank mRNA | AK052404.1 | - | 15 - 16 | 2030 | 61 | |
AK054305.1 | - | 15 - 16 | 2031 | 61 | ||
AK077847.1 | - | 15 - 16 | 2050 | 61 | ||
AK090134.1 | - | 15 - 16 | 2031 | 61 | ||
AK161161.1 | - | 2 - 3 | 157 | 61 | ||
AK164804.1 | - | 15 - 16 | 2035 | 61 | ||
BC139002.1 | - | 15 - 16 | 2030 | 8862024 | 61 | |
BC145492.1 | - | 15 - 16 | 2030 | 9054055 | 61 | |
X80169.1 | - | 15 - 16 | 2034 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
