Assay Details
| Gene Symbol: Snap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291056.1 | NP_001277985.1 | 2 - 3 | 281 | 73 | |
| NM_011428.3 | NP_035558.1 | 2 - 3 | 281 | 73 | ||
| GenBank mRNA | AF483516.1 | - | 1 - 2 | 68 | 73 | |
| AF483517.1 | - | 1 - 2 | 68 | 73 | ||
| AK078038.1 | - | 2 - 3 | 279 | 73 | ||
| AK138653.1 | - | 2 - 3 | 249 | 73 | ||
| AU080862.1 | - | 2 - 3 | 248 | 73 | ||
| BC018249.1 | - | 2 - 3 | 169 | 4504644 | 73 | |
| CB232574.1 | - | 2 - 3 | 284 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snap25
Genomic Map