Assay Details
| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159593.1 | NP_001153065.1 | 3 - 4 | 978 | 62 | |
| NM_015747.2 | NP_056562.1 | 3 - 4 | 978 | 62 | ||
| GenBank mRNA | AK035898.1 | - | 3 - 4 | 949 | 62 | |
| AK084159.1 | - | 3 - 4 | 951 | 62 | ||
| AK155506.1 | - | 3 - 4 | 951 | 62 | ||
| BC015085.1 | - | 3 - 4 | 878 | 4015575 | 62 | |
| M73696.1 | - | 2 - 3 | 897 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc20a1
Genomic Map