Assay Details
| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015747.2 | NP_056562.1 | 5 - 6 | 1165 | 82 | |
| GenBank mRNA | AK155506.1 | - | 5 - 6 | 1138 | 82 | |
| BC015085.1 | - | 5 - 6 | 1065 | 4015575 | 82 | |
| M73696.1 | - | 4 - 5 | 1084 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc20a1
Genomic Map