Assay Details
| Gene Symbol: Il1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008361.3 | NP_032387.1 | 1 - 2 | 55 | 63 | |
| GenBank mRNA | AK156396.1 | - | 1 - 2 | 75 | 63 | |
| AK157245.1 | - | 1 - 2 | 75 | 63 | ||
| AK168047.1 | - | 1 - 2 | 75 | 63 | ||
| AK224980.1 | - | 1 - 2 | 75 | 63 | ||
| AK225002.1 | - | 1 - 2 | 74 | 63 | ||
| BC011437.1 | - | 1 - 2 | 55 | 3989461 | 63 | |
| M15131.1 | - | 1 - 2 | 64 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK168047.1, BC011437.1, M15131.1, NM_008361.3 transcripts.
Set Membership: |
|

Il1b
Genomic Map