Assay Details
| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028248.2 | NP_082524.2 | 2 - 3 | 263 | 109 | |
| GenBank mRNA | AK011959.1 | - | 2 - 3 | 184 | 109 | |
| AK050203.1 | - | 2 - 3 | 255 | 109 | ||
| AK164504.1 | - | 2 - 3 | 245 | 109 | ||
| AK166991.1 | - | 2 - 3 | 243 | 109 | ||
| AK171811.1 | - | 3 - 4 | 179 | 109 | ||
| AK172611.1 | - | 3 - 4 | 179 | 109 | ||
| BC031488.1 | - | 2 - 3 | 219 | 109 | ||
| CA479832.1 | - | 2 - 3 | 485 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011959.1, AK171811.1, AK172611.1 transcripts.
Set Membership: |
|

Tmem87b
Genomic Map