Assay Details
| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028248.2 | NP_082524.2 | 3 - 4 | 360 | 74 | |
| GenBank mRNA | AK011959.1 | - | 3 - 4 | 281 | 74 | |
| AK050203.1 | - | 3 - 4 | 352 | 74 | ||
| AK164504.1 | - | 3 - 4 | 342 | 74 | ||
| AK166991.1 | - | 3 - 4 | 340 | 74 | ||
| AK171811.1 | - | 4 - 5 | 276 | 74 | ||
| AK172611.1 | - | 4 - 5 | 276 | 74 | ||
| BC031488.1 | - | 3 - 4 | 316 | 74 | ||
| CA479832.1 | - | 3 - 4 | 582 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem87b
Genomic Map