Assay Details
| Gene Symbol: Tmem87b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028248.2 | NP_082524.2 | 9 - 10 | 973 | 67 | |
| GenBank mRNA | AK011959.1 | - | 9 - 10 | 894 | 67 | |
| AK050203.1 | - | 9 - 10 | 965 | 67 | ||
| AK171811.1 | - | 10 - 11 | 889 | 67 | ||
| AK172611.1 | - | 10 - 11 | 889 | 67 | ||
| BC031488.1 | - | 9 - 10 | 929 | 67 | ||
| CN458042.1 | - | 1 - 2 | 108 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem87b
Genomic Map