Assay Details
| Gene Symbol: Polr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009086.2 | NP_033112.2 | 2 - 3 | 434 | 59 | |
| GenBank mRNA | AK052672.1 | - | 2 - 3 | 404 | 59 | |
| AK083732.1 | - | 2 - 3 | 416 | 59 | ||
| AK143501.1 | - | 2 - 3 | 371 | 59 | ||
| AK144616.1 | - | 2 - 3 | 421 | 59 | ||
| AK156712.1 | - | 2 - 3 | 425 | 59 | ||
| BC060656.1 | - | 2 - 3 | 364 | 30355130 | 59 | |
| U58280.1 | - | 2 - 3 | 398 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr1b
Genomic Map