Assay Details
| Gene Symbol: Polr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009086.2 | NP_033112.2 | 8 - 9 | 1423 | 60 | |
| GenBank mRNA | AK052672.1 | - | 8 - 9 | 1393 | 60 | |
| AK083732.1 | - | 8 - 9 | 1405 | 60 | ||
| AK143501.1 | - | 8 - 9 | 1360 | 60 | ||
| AK144616.1 | - | 8 - 9 | 1410 | 60 | ||
| AK156712.1 | - | 8 - 9 | 1414 | 60 | ||
| BC060656.1 | - | 8 - 9 | 1353 | 30355130 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polr1b
Genomic Map